Please login to your online account to display your discounted pricing

Methanol - D4, deuteration degree min. 99.95% for NMR spectroscopy MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$249.77 - $375.59

Chemical Identifiers

Chemical Name or Material Methanol - D4
CAS 811-98-3
Physical Form Liquid
Assay Percent Range 99.95% min.
Molecular Formula CD3OD
Formula Weight 36.07
Synonym Tetradeuteromethanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1060250005

 
emd millipore corporation
1060250005
Glass ampoule 10 x 0.5mL Each for $249.77

M1060250009

 
emd millipore corporation
1060250009
Glass ampoule 10 x 0.75mL Each for $375.59
Description & Specifications

Specifications

Chemical Name or Material Methanol - D4
CAS 811-98-3
Odor Characteristic
Packaging Glass ampoule
Ignition Point 455°C (851°F)
Vapor Pressure 128 hPa at 20°C (68°F)
Viscosity 0.597mPa&middnot;s (20°C/68°F)
Physical Form Liquid
Quantity 10 x 0.5mL
Assay Percent Range 99.95% min.
Color Colorless
Molecular Formula CD3OD
Formula Weight 36.07
Synonym Tetradeuteromethanol