missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol - D4, deuteration degree min. 99.95% for NMR spectroscopy MagniSolv™, MilliporeSigma™

For NMR Spectroscopy

Manufacturer:  MilliporeSigma 1060250009

 View more versions of this product

Catalog No. M1060250009



Specifications

Specifications

Methanol - D4
Characteristic
128 hPa at 20°C (68°F)
Colorless
811-98-3
CH4O
OKKJLVBELUTLKV-MZCSYVLQSA-N
trideuterio(deuteriooxy)methane
71568
Liquid
455°C (851°F)
Glass ampoule
0.597mPa·s (20°C/68°F)
10 x 0.75mL
99.95% min.
methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
CO
36.066
36.07

Chemical Identifiers

811-98-3
36.066
methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
trideuterio(deuteriooxy)methane
CH4O
OKKJLVBELUTLKV-MZCSYVLQSA-N
71568
CO
Documents
Promotions

Promotions