Please login to your online account to display your discounted pricing

Methanol - D4, deuteration degree min. 99.8 % for NMR spectroscopy MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$30.31 - $1,182.89

Chemical Identifiers

Chemical Name or Material Methanol - d4
CAS 811-98-3
Physical Form Liquid
Assay Percent Range 99.8% min.
Molecular Formula CD3OD
Formula Weight 36.07
Synonym Tetradeuteromethanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1060280001

 
emd millipore corporation
1060280001
Glass ampoule 1mL Each for $30.31

M1060280100

 
emd millipore corporation
1060280100
Glass Bottle 100mL Each for $1,182.89

M1060280009

 
emd millipore corporation
1060280009
Glass ampoule 10 x 0.75mL Each for $181.99

M1060280010

 
emd millipore corporation
1060280010
Septum ampoule 10mL Each for $220.73

M1060280005

 
emd millipore corporation
1060280005
Glass ampoule 10 x 0.5mL Each for $121.17

M1060280025

 
emd millipore corporation
1060280025
Glass Bottle 25mL Each for $398.69
Description & Specifications

Specifications

Formula Weight 36.07
Quantity 1mL
Color Colorless
UN Number 1230
Viscosity 0.597mPa&middnot;s (20°C/68°F)
Physical Form Liquid
Vapor Pressure 128 hPa at 20°C (68°F)
CAS 811-98-3
Odor Characteristic
Packaging Glass ampoule
Chemical Name or Material Methanol - D4
Assay Percent Range 99.8% min.
Molecular Formula CD3OD
Synonym Tetradeuteromethanol
Ignition Point 455°C (851°F)