Please login to your online account to display your discounted pricing

Alfa Aesar™ Methanol-d4, 100%(Isotopic)

Chemical Identifiers

Chemical Name or Material Methanol-d4
CAS 811-98-3
Name Note Isotopic
Formula Weight 36.07
MDL Number MFCD00044637
UN Number UN1230
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42320KV

  SDS
alfa aesar™
42320KV
10 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 36.07
Packaging Prescored Ampules
Chemical Name or Material Methanol-d4
TSCA TSCA
Density 0.89g/mL
Name Note Isotopic
Boiling Point 64.7°C
Melting Point −98°C
Physical Form Liquid
Assay 100%
Quantity 10 each
Color Colorless
Flash Point 11°C (52°F)
MDL Number MFCD00044637
CAS 811-98-3
UN Number UN1230
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H300-H310-H330-H370
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs.
Odor Mild
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHANOL
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H300-H310-H330-H370
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs.