Please login to your online account to display your discounted pricing

Methanol-d3, for NMR, 99.5% atom D, ACROS Organics™

$161.49 - $161.49

Chemical Identifiers

Chemical Name or Material Methanol-d3
Name Note 99.5 atom % D, For NMR
CAS 1849-29-2
Purity Grade For NMR
Molecular Formula CHD3O
Linear Formula CD3OH
Formula Weight 35.07
MDL Number MFCD00055877
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC300730025

  SDS
thermo fisher scientific
300730025
Glass bottle 2.5mL Each for $161.49
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 35.07
Quantity 2.5mL
Color Clear colorless
Density 0.8670g/mL
Purity Grade For NMR
Chemical Name or Material Methanol-d3
Name Note 99.5 atom % D, For NMR
Molecular Formula CHD3O
CAS 1849-29-2
Packaging Glass bottle
Water 0.1% max. (Coulometric)
Linear Formula CD3OH
MDL Number MFCD00055877
Deuterium (D) 99.4% min.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes damage to organs.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For NMR

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes damage to organs.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.