Please login to your online account to display your discounted pricing

Methanol-D3, deuteration degree min. 99.5% for NMR spectroscopy, MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$89.44 - $355.21

Chemical Identifiers

Chemical Name or Material Methanol-D3
CAS 1849-29-2
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula CD3OH
Formula Weight 35.06
Synonym Tetradeuteromethanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M8150520005

 
emd millipore corporation
8150520005
Glass ampoule 5mL Each for $355.21

M8150520001

 
emd millipore corporation
8150520001
Glass ampoule 1mL Each for $89.44
Description & Specifications

Specifications

Formula Weight 35.06
Quantity 5mL
Color Colorless
Viscosity 0.597mPa&middnot;s (20°C/68°F)
Physical Form Liquid
Molecular Formula CD3OH
CAS 1849-29-2
Odor Characteristic
Packaging Glass ampoule
Chemical Name or Material Methanol-D3
Vapor Pressure 128hPa (20°C)
Assay Percent Range 99.5% min.
Synonym Tetradeuteromethanol
Ignition Point 455°C