Please login to your online account to display your discounted pricing

Methanol-D3, deuteration degree min. 99.5% for NMR spectroscopy, MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Quantity 5mL
Packaging Glass ampoule

Manufacturer: emd millipore corporation  8150520005

 View more versions of this product

Catalog No. M8150520005

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 35.06
Quantity 5mL
Color Colorless
Viscosity 0.597mPa&middnot;s (20°C/68°F)
Physical Form Liquid
Molecular Formula CD3OH
CAS 1849-29-2
Odor Characteristic
Packaging Glass ampoule
Chemical Name or Material Methanol-D3
Vapor Pressure 128hPa (20°C)
Assay Percent Range 99.5% min.
Synonym Tetradeuteromethanol
Ignition Point 455°C

Specifications

Chemical Name or Material Methanol-D3
CAS 1849-29-2
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula CD3OH
Formula Weight 35.06
Synonym Tetradeuteromethanol