missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™

Product filtered through a 0.2µm filter

Manufacturer:  Honeywell-Chromasolv 348606X1L

Catalog No. 6000944



Description

Description

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
Specifications

Specifications

Methanol
67-56-1
CH3OH
1098229
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04g/mol
CHROMASOLV™
64.7°C
-98°C
n20/D 1.329
97.68mmHg (20°C)
for HPLC
CH4O
MFCD00004595
carbinol, colonial spirit, columbian spirit, methyl alcohol, methyl hydroxide, methylol, pyroxylic spirit, wood alcohol, wood naphtha, wood spirit
CO
32.04
CHEBI:17790
≥99.9%
0.791g/mL at 25°C
Undesignated
6 x 1L
1.11 (vs. air)
Safety and Handling

Safety and Handling

P210-P260-P280-P301 + P310-P311

H225-H301 + H311 + H331-H370

EINECSNumber : 200-659-6

RTECSNumber : PC1400000

SDS
Documents
Product Certifications
Promotions

Promotions