missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, CHROMASOLV™, Honeywell Riedel-de Haën™
CHROMASOLV™, for GC/MS analysis of volatile organics,≥99.9%
$2402.38 - $2402.38
Chemical Identifiers
| CAS | 67-56-1 |
|---|---|
| Molecular Formula | CH4O |
| Molecular Weight (g/mol) | 32.04 |
| MDL Number | MFCD00004595 |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| PubChem CID | 887 |
| ChEBI | CHEBI:17790 |
| IUPAC Name | methanol |
| SMILES | CO |
Description
Honeywell Chromasolv™ GC-headspace solvents:
- developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
- very pure solvents with extremely low concentrations of the defined residual solvents
- state-of-the art production and application tests
Chemical Identifiers
| 67-56-1 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| CHEBI:17790 | |
| CO |
| CH4O | |
| MFCD00004595 | |
| 887 | |
| methanol |
Specifications
| 67-56-1 | |
| 6 x 1 L | |
| 9°C (48.2°F) | |
| Glass Bottle | |
| CH3OH | |
| 410 mmHg (50°C), 97.68 mmHg (20°C) | |
| 1098229 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 887 | |
| 32.04g/mol | |
| CHROMASOLV™ |
| 0.791g/mL at 25°C, 0.791g/mL at 25°C | |
| 0.36 | |
| 725°F | |
| CH4O | |
| MFCD00004595 | |
| UN1230 | |
| methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha | |
| CO | |
| 32.04 | |
| CHEBI:17790 | |
| ≥99.9% (GC) | |
| Methanol |
Safety and Handling
P210-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378-P403 + P235
RTECSNumber : PC1400000