missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, Certified, 50% (v/v), LabChem™

$77.63 - $140.43

Chemical Identifiers

CAS 7732-18-5,67-56-1
CAS Max % 55.5,44.5
Linear Formula CH3OH
Formula Weight 32.04
UN Number UN1993
Synonym Wood Alcohol
Chemical Name or Material Methanol
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
LC168302 Lab Chem
LC168302
Poly Bottle 1L Each for $77.63
LC168304 Lab Chem
LC168304
Poly Bottle 4L Each for $140.43
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,67-56-1
CH3OH
UN1993
Methanol
55.5,44.5
32.04
Wood Alcohol
Certified Grade

Specifications

7732-18-5,67-56-1
Liquid
32.04
Wood Alcohol
Poly Bottle
0.928 ±0.008
27°C
Carbon monoxide; Carbon dioxide; May release flammable gases
1L
50% (v/v)
55.5,44.5
CH3OH
UN1993
Passes Test
Soluble in water
Room Temperature
1.96cSt
Clear, colorless
Methanol
Certified Grade

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed or in contact with skin.
Harmful if inhaled.
Causes damage to organs (central nervous system, optic nerve).

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
In case of fire: Use alcohol resistant foam, sand, extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

Danger