Please login to your online account to display your discounted pricing

Methanol, BAKER ANALYZED™ ULTRA LC/MS, J.T.Baker™

For cutting-edge LC/MS applications

$273.44 - $448.41

Chemical Identifiers

Chemical Name or Material Methanol
CAS 67-56-1
Purity Grade BAKER ANALYZED™ ULTRA LC/MS
Molecular Formula CH3OH
Formula Weight 32.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003340

 
avantor performance
986302
Narrow Mouth Flint 1L Each for $273.44

02003339

 
avantor performance
986301
Narrow Mouth Flint 1L Case of 2 Each for $448.41
Description & Specifications

Specifications

Melting Point 113° to 116°C
Formula Weight 32.04
Chemical Name or Material Methanol
Packaging Narrow Mouth Flint
Specific Gravity 1L = 0.79kg
Purity Grade BAKER ANALYZED™ ULTRA LC/MS
Flash Point 12.00°C
Quantity 1L
Molecular Formula CH3OH
CAS 67-56-1

    For Use in LC/MS and UHPLC; For Laboratory, Research, or Manufacturing Use