Learn More
Methanol, ACS, absolute, low acetone, 99.8+%, Thermo Scientific Chemicals
$49.29 - $185.88
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA31721AP
|
Thermo Scientific Chemicals
031721AP |
500 mL |
Each for $49.29
|
|
|||||
AA31721K2
|
Thermo Scientific Chemicals
031721K2 |
1 L |
Each for $62.42
|
|
|||||
AA31721M1
|
Thermo Scientific Chemicals
031721M1 |
4 x 1 L |
Each for $185.88
|
|
|||||
AA31721K7
|
Thermo Scientific Chemicals
031721K7 |
4 L |
Each for $158.00
|
|
|||||
Description
Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role for the measurement of methyl ester content of pectin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
°C to 98°C | |
Colorless | |
0.791 | |
500 mL | |
Mild | |
1.329 | |
CH4O | |
128 hPa (96mm Hg) at 20°C | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 | |
Methanol |
67-56-1 | |
Colorless liquid | |
64.7°C | |
11°C (52°F) | |
Liquid | |
>99.8% | |
MFCD00004595 | |
1098229 | |
Miscible with water and most of the organic solvents including ethyl acetate,toluene,dichloromethane,chloroform,tetrahydrofuran,and acetone. | |
CO | |
32.04 | |
CHEBI:17790 | |
ACS |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
H225-H301+H311+H331-H370-H372
DOTInformation : DOT Class: 3; Packing Group: II
EINECSNumber : 200-659-6
RTECSNumber : PC1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only