missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, ACS, absolute, low acetone, >99.8%, Alfa Aesar™

Manufacturer:  Alfa Aesar 31721AP

 View more versions of this product

Catalog No. AA31721AP



Specifications

Specifications

Methanol
Colorless liquid
CH4O
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.042
CHEBI:17790
Liquid
Mild
Colorless
67-56-1
>99.8%
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
ACS
128 hPa (96mm Hg) at 20°C
500mL

Chemical Identifiers

67-56-1
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO
Safety and Handling

Safety and Handling

H225-H301-H311-H331-H370: Highly flammable liquid and vapour. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes damage to organs.

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 200-659-6

SDS Documents