missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, ACS, absolute, low acetone, >99.8%, Alfa Aesar™

Manufacturer:  Alfa Aesar 31721K7

 View more versions of this product

Catalog No. AA31721K7



Specifications

Specifications

Methanol
ACS
CH4O
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.042
CHEBI:17790
Liquid
4L
67-56-1
>99.8%
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
Colorless

Chemical Identifiers

67-56-1
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO
Safety and Handling

Safety and Handling

H225-H301-H311-H331-H370: Highly flammable liquid and vapour. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes damage to organs.

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 200-659-6

SDS Documents