Please login to your online account to display your discounted pricing

Methanol, Absolute, PHOTREX™ Reagent, J.T.Baker™


High purity solvents characterized for use in UV, visible and IR spectrophotometry

Quantity 4L
Packaging Narrow Mouth Amber Glass Bottle

Manufacturer: avantor performance  906903

 View more versions of this product

Catalog No. 02003353

  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 32.04
CAS 67-56-1
Chemical Name or Material Methanol
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.80kg
Name Note Absolute
Purity Grade PHOTREX™ Reagent
Flash Point 12.00°C
Quantity 4L
Molecular Formula CH3OH

Specifications

Chemical Name or Material Methanol
CAS 67-56-1
Name Note Absolute
Purity Grade PHOTREX™ Reagent
Molecular Formula CH3OH
Formula Weight 32.04

    For Spectrophotometry