Please login to your online account to display your discounted pricing

Methanol, Absolute, PHOTREX™ Reagent, J.T.Baker™

High purity solvents characterized for use in UV, visible and IR spectrophotometry

$704.10 - $704.10

Chemical Identifiers

Chemical Name or Material Methanol
CAS 67-56-1
Name Note Absolute
Purity Grade PHOTREX™ Reagent
Molecular Formula CH3OH
Formula Weight 32.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003353

 
avantor performance
906903
Narrow Mouth Amber Glass Bottle 4L Case of 4 Each for $704.10
Description & Specifications

Specifications

Formula Weight 32.04
CAS 67-56-1
Molecular Formula CH3OH
Name Note Absolute
Quantity 4L
Flash Point 12.00°C
Specific Gravity 1L = 0.80kg
Chemical Name or Material Methanol
Packaging Narrow Mouth Amber Glass Bottle
Purity Grade PHOTREX™ Reagent

    For Spectrophotometry