Please login to your online account to display your discounted pricing

Methanol, 99.8%, for residue analysis, ECD tested for pesticide analysis, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  326630025

 View more versions of this product

Catalog No. AC326630025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.8%
Physical Form Liquid
Formula Weight 32.04
Quantity 2.5L
Color Clear
Beilstein 01, IV, 1227
Fieser 11,322; 13,337
Merck Index 15, 6029
TSCA TSCA
RTECS Number PC1400000
EINECS Number 200-659-6
Purity Grade For residue analysis
Density 0.7910g/mL
Chemical Name or Material Methanol
Additional Information GC/ECD (as lindane): max 5ng/L
Packaging Glass bottle
Residue after Evaporation 0.0002% max.
Water 0.1% max. (Coulometric)
Molecular Formula CH4O
CAS 67-56-1
Synonym Methyl alcohol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Acetic Acid 0.003% max.
Name Note ECD Tested, for Pesticide Analysis
Assay Percent Range 99.8%
Linear Formula CH3OH
MDL Number MFCD00004595

Specifications

Chemical Name or Material Methanol
Name Note ECD Tested, for Pesticide Analysis
CAS 67-56-1
Purity Grade For residue analysis
Assay ≥99.8%
Assay Percent Range 99.8%
Molecular Formula CH4O
Linear Formula CH3OH
Formula Weight 32.04
MDL Number MFCD00004595
Synonym Methyl alcohol

For residue analysis

Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.

SDS & Safety Documents