Please login to your online account to display your discounted pricing

Methanol, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™


Quantity 1L
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  364390010

 View more versions of this product

Catalog No. AC364390010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.8% (GC)
Boiling Point 64.7°C
Purity Grade Extra Dry over Molecular Sieve
Melting Point −98°C
Physical Form Liquid
Formula Weight 32.04
Quantity 1L
Color Clear colorless
Beilstein 01, IV, 1227
Fieser 11,322; 13,337
Merck Index 15, 6029
Flash Point 12°C (53.6°F)
Packaging AcroSeal™ Glass Bottle
Density 0.7910g/mL
Water 0.005% Max. (Coulometric)
Synonym Methyl alcohol
Assay Percent Range 99.8%
Chemical Name or Material Methanol
Molecular Formula CH4O
Name Note water <50 ppm
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00004595
Acidity Alkalinity 0.0005 meq/g max.
Infrared Spectrum Authentic
Linear Formula CH3OH
Acidity 0.003% max. (as HCOOH)
Additional Information Vapor Pressure: 128hPa at 20°C
Health Hazard 1 Danger
CAS 67-56-1

Specifications

Chemical Name or Material Methanol
Name Note water <50 ppm
CAS 67-56-1
Purity Grade Extra Dry over Molecular Sieve
Assay ≥99.8% (GC)
Assay Percent Range 99.8%
Molecular Formula CH4O
Linear Formula CH3OH
Formula Weight 32.04
MDL Number MFCD00004595
Synonym Methyl alcohol
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.

SDS & Safety Documents