Please login to your online account to display your discounted pricing

Methanol, 99.8%, Extra Dry, AcroSeal™, ACROS Organics™

$73.41 - $73.41

Chemical Identifiers

Chemical Name or Material Methanol
CAS 67-56-1
Purity Grade Extra Dry
Assay ≥99.8%
Assay Percent Range 99.8%
Molecular Formula CH4O
Linear Formula CH3OH
Formula Weight 32.04
MDL Number MFCD00004595
Synonym Methyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC610400010

  SDS
thermo fisher scientific
610400010
Amber Glass Bottle 1L Each for $73.41
Description & Specifications

Specifications

Assay ≥99.8%
Melting Point −98°C
Formula Weight 32.04
Quantity 1L
Molecular Formula CH4O
UN Number 1230
Packaging Amber Glass Bottle
Flash Point 64.7°C (148.46°F)
EINECS Number 200-659-6
TSCA TSCA
Purity Grade Extra Dry
Density 0.7910g/mL
RTECS Number PC1400000
Beilstein 01, IV, 1227
Fieser 11,322; 13,337
Merck Index 15, 6029
Chemical Name or Material Methanol
Identification Pass Test
Additional Information Vapor Pressure: 128hPa at 20°C
Water 50ppm max.
MDL Number MFCD00004595
CAS 67-56-1
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.
Assay Percent Range 99.8%
Linear Formula CH3OH
Synonym Methyl alcohol
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.