Learn More
Methanol, 99.8%, for electronic use, residue free, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 327900010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For electronic useSpecifications
Methanol | |
Residue Free | |
0.7910g/mL | |
1 L | |
Glass Bottle | |
1.3280 to 1.3300 | |
99.8% | |
CH4O | |
MFCD00004595 | |
0.1% max. (Coulometric) | |
11,322; 13,337 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 | |
Electronic |
-98.0°C | |
67-56-1 | |
64.7°C | |
12°C | |
Liquid | |
0.0005% max. | |
0.791 | |
CH3OH | |
0.55 mPa.s (20°C) | |
01, IV, 1227 | |
15, 6029 | |
Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents | |
CO | |
32.04 | |
CHEBI:17790 | |
99.8% |
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 200-659-6
RTECSNumber : PC1400000
TSCA : TSCA
RUO – Research Use Only