missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, 99.8+%, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 423955000

 View more versions of this product

Catalog No. AC423955000




Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methanol
67-56-1
64.7°C
12°C
Liquid
0.001% max.
0.791
CH4O
CH3OH
Vapor Pressure: 128hPa at 20°C
0.1% max. (K.F.)
11,322; 13,337
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
ACS Reagent
-98.0°C
0.7910g/mL
500 mL
EcoSafPak™ Glass Bottle
1.3280 to 1.3300
99.8+%
0.0003 meq/g max.
0.0002 meq/g max.
MFCD00004595
0.55 mPa.s (20°C)
01, IV, 1227
15, 6029
Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
CO
32.04
CHEBI:17790
≥99.8%

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only