missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methanesulfonyl fluoride, 98%

Manufacturer:  Alfa Aesar™ H5346014

 View more versions of this product

Catalog No. AAH5346014



Specifications

Specifications

Methanesulfonyl fluoride
1.36
25g
CH3FO2S
UN3389
KNWQLFOXPQZGPX-UHFFFAOYSA-N
methanesulfonyl fluoride
11207
98%
1.427
123°C to 124°C
558-25-8
MFCD00007423
ch3so2f
CS(=O)(=O)F
98.091
98.09

Chemical Identifiers

558-25-8
98.091
KNWQLFOXPQZGPX-UHFFFAOYSA-N
11207
CS(=O)(=O)F
CH3FO2S
MFCD00007423
ch3so2f
methanesulfonyl fluoride
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H314-H318
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H300+H310+H330-H314

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 209-192-2

RTECSNumber : PB2975000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents