missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanesulfonic Acid 99.0+%, TCI America™

Manufacturer:  TCI America M009325G

 View more versions of this product

Catalog No. M009325G



Specifications

Specifications

Methanesulfonic Acid
Yellow
75-75-2
MFCD00007518
ccris 2783
CS(=O)(=O)O
96.1
CHEBI:27376
Clear Liquid at 20°C
122°C
25g
CH4O3S
2586
AFVFQIVMOAPDHO-UHFFFAOYSA-N
methanesulfonic acid
6395
96.10
≥99.0% (T)

Chemical Identifiers

75-75-2
96.1
AFVFQIVMOAPDHO-UHFFFAOYSA-N
6395
methanesulfonic acid
CH4O3S
MFCD00007518
ccris 2783
CHEBI:27376
CS(=O)(=O)O
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1582

RTECSNumber : PB1140000

TSCA : Yes

Documents
Promotions

Promotions