missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanesulfonic Acid Solution, Honeywell™ Fluka™

$98.86 - $4,025.05

Chemical Identifiers

CAS 75-75-2
Molecular Formula CH4O3S
Molecular Weight (g/mol) 96.1
MDL Number MFCD00007518
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
Synonym ccris 2783
PubChem CID 6395
ChEBI CHEBI:27376
IUPAC Name methanesulfonic acid
SMILES CS(=O)(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044955 View Documents Honeywell Chemicals
1783410ML
10mL Glass Bottle Each for $98.86
 
60044954 View Documents Honeywell Chemicals
17834100ML
100mL Glass Bottle Each for $716.67
 
60044956 View Documents Honeywell Chemicals
178341L
1L Glass Bottle Each for $4,025.05
 
Specifications

Chemical Identifiers

75-75-2
96.1
AFVFQIVMOAPDHO-UHFFFAOYSA-N
6395
methanesulfonic acid
CH4O3S
MFCD00007518
ccris 2783
CHEBI:27376
CS(=O)(=O)O

Specifications

75-75-2
CH3SO3H
UN3265
ccris 2783
CS(=O)(=O)O
96.1
CHEBI:27376
approx. 1M in water
Glass Bottle
Colorless
Methanesulfonic Acid Solution, For ion chromatography
CH4O3S
MFCD00007518
1446024
AFVFQIVMOAPDHO-UHFFFAOYSA-N
methanesulfonic acid
6395
96.11g/mol
1.039g/mL at 20°C
Liquid
10mL
Safety and Handling

Safety and Handling

P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

H290-H314

DOTInformation : 8/PGII