Please login to your online account to display your discounted pricing

Methanesulfonic Acid, BAKER™, J.T.Baker™

A grade of chemicals for laboratory use

$180.68 - $1,376.68

Chemical Identifiers

Chemical Name or Material Methanesulfonic Acid
CAS 75-75-2
Molecular Formula CH3SO3H
Formula Weight 96.1
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

02003436

 
avantor performance
P68407
Each for $180.68

02003437

 
avantor performance
P68409
Each for $1,376.68
Description & Specifications

Specifications

Formula Weight 96.1
Chemical Name or Material Methanesulfonic Acid
CAS 75-75-2
Molecular Formula CH3SO3H
Quantity 500g
Specific Gravity 1L = 1.48kg
Flash Point 189.00°C
Packaging Narrow Mouth Amber Glass Bottle