Learn More
Methanesulfonic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 432970010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methanesulfonic acid | |
19.0°C | |
1.4810g/mL | |
189°C | |
98% | |
CH4O3S | |
CH3SO3H | |
1.481 | |
methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane | |
AFVFQIVMOAPDHO-UHFFFAOYSA-N | |
methanesulfonic acid | |
6395 | |
CHEBI:27376 | |
98% |
75-75-2 | |
Yellow to Brown | |
167.0°C (10.0 mmHg) | |
Authentic | |
Glass Bottle | |
1.4252 to 1.4315 | |
1 L | |
15,6026 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and ethanol, insoluble in hexane and methylcyclopentane, 1.50wt% benzene -0.23wt% 0-chlorotoluene (26-28°C), 0.38wt% toluene -0.47wt% ethyl disulfide (26-28°C) | |
CS(=O)(=O)O | |
96.1 | |
11.6 mPa.s (25°C) | |
96.1 | |
Liquid |
Chemical Identifiers
75-75-2 | |
96.1 | |
methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane | |
CHEBI:27376 | |
CS(=O)(=O)O |
CH4O3S | |
AFVFQIVMOAPDHO-UHFFFAOYSA-N | |
6395 | |
methanesulfonic acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
May be corrosive to metals.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
D
GHS Signal Word: Danger
EINECSNumber : 200-898-6
RUO – Research Use Only