Learn More
Methanesulfonamide, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1088509
Description
Methanesulfonamide will react with thionyl chloride, cyanates, carbon disulfide and ketones and aldehydes to prepare pharmaceuticals, brightening agent and other target molecules. It is also used in biological studies to predict binding affinity and binding mode of protein ligand complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethanesulfonamide will react with thionyl chloride, cyanates, carbon disulfide and ketones and aldehydes to prepare pharmaceuticals, brightening agent and other target molecules. It is also used in biological studies to predict binding affinity and binding mode of protein ligand complexes.
Solubility
Slightly soluble in DMSO and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Methanesulfonamide | |
87°C to 93°C | |
MFCD00007940 | |
1740835 | |
Slightly soluble in DMSO and methanol. | |
CS(N)(=O)=O | |
95.12 | |
95.13 |
3144-09-0 | |
CH5NO2S | |
10 g | |
methylsulfonamide, methane sulfonamide, methanesulphonamide, methansulfonamid, methyl sulfonamide, aminosulfane dioxide, methyl sulfonylamine, ch3so2nh2, mesylamine, methansulfonamide | |
HNQIVZYLYMDVSB-UHFFFAOYSA-N | |
methanesulfonamide | |
72879 | |
≥98% |
Chemical Identifiers
3144-09-0 | |
95.12 | |
HNQIVZYLYMDVSB-UHFFFAOYSA-N | |
72879 | |
CS(N)(=O)=O |
CH5NO2S | |
MFCD00007940 | |
methylsulfonamide, methane sulfonamide, methanesulphonamide, methansulfonamid, methyl sulfonamide, aminosulfane dioxide, methyl sulfonylamine, ch3so2nh2, mesylamine, methansulfonamide | |
methanesulfonamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 221-553-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only