missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methanesulfinic acid sodium salt, 95%

Manufacturer:  Alfa Aesar™ L1136014

 View more versions of this product

Catalog No. AAL1136014



Description

Description

Applications
Methanesulfinic acid sodium salt is used in the preparation of alkyl methyl sulfone and methyl bis(4-tolyl)sulfoniumtrifluoromethanesulfonate. It is also employed in the synthesis of two sulfonyl type reversible addition-fragmentation transfer (RAFT) agents such as benzyl methylsulfonyldithioformate and benzyl phenylsulfonyldithioformate.

Solubility
Soluble in water.

Notes
Air sensitive. Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

Methanesulfinic acid sodium salt
25g
CH3NaO2S
MFCD00040392
sodium methanesulfinate, sodium methanesulphinate, methanesulfinic acid sodium salt, sodium methylsulfinate, methanesulfinic acid, sodium salt, sodium methane sulfinate, methanesulfinicacidsodiumsalt, methanesulphinic acid sodium salt, methane sulfinic acid sodium salt, meso2na
CS(=O)[O-].[Na+]
102.083
102.09
227°C to 230°C
20277-69-4
CH3SO2Na
3565430
LYPGDCWPTHTUDO-UHFFFAOYSA-M
sodium;methanesulfinate
2733271
95%

Chemical Identifiers

20277-69-4
102.083
LYPGDCWPTHTUDO-UHFFFAOYSA-M
2733271
CS(=O)[O-].[Na+]
CH3NaO2S
MFCD00040392
sodium methanesulfinate, sodium methanesulphinate, methanesulfinic acid sodium salt, sodium methylsulfinate, methanesulfinic acid, sodium salt, sodium methane sulfinate, methanesulfinicacidsodiumsalt, methanesulphinic acid sodium salt, methane sulfinic acid sodium salt, meso2na
sodium;methanesulfinate
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents