missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methanesulfinic acid, sodium salt, 85%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 392960500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methanesulfinic acid, sodium salt | |
Authentic | |
Glass bottle | |
CH3SO2Na | |
sodium methanesulfinate, sodium methanesulphinate, methanesulfinic acid sodium salt, sodium methylsulfinate, methanesulfinic acid, sodium salt, sodium methane sulfinate, methanesulfinicacidsodiumsalt, methanesulphinic acid sodium salt, methane sulfinic acid sodium salt, meso2na | |
LYPGDCWPTHTUDO-UHFFFAOYSA-M | |
sodium;methanesulfinate | |
2733271 | |
85% |
20277-69-4 | |
85% | |
CH3NaO2S | |
50g | |
Solubility in water: soluble | |
CS(=O)[O-].[Na+] | |
102.09 | |
102.09 |
Chemical Identifiers
20277-69-4 | |
102.09 | |
sodium methanesulfinate, sodium methanesulphinate, methanesulfinic acid sodium salt, sodium methylsulfinate, methanesulfinic acid, sodium salt, sodium methane sulfinate, methanesulfinicacidsodiumsalt, methanesulphinic acid sodium salt, methane sulfinic acid sodium salt, meso2na | |
sodium;methanesulfinate |
CH3NaO2S | |
LYPGDCWPTHTUDO-UHFFFAOYSA-M | |
2733271 | |
CS(=O)[O-].[Na+] |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 243-669-6
RUO â Research Use Only