missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methallyl phenyl ether, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2236906

 View more versions of this product

Catalog No. AAB2236906



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methallyl phenyl ether
0.964
77°C (171°F)
1.515
5 g
methallyl phenyl ether, 2-methylallyloxy benzene, ether, 2-methylallyl phenyl, benzene, 2-methyl-2-propenyl oxy, 2-methylallyl phenyl ether, 2-methylprop-2-en-1-yl oxy benzene, 2-methyl-2-propen-1-yl oxy benzene, phenyl-methallyl ether, acmc-1b20w
CC(=C)COC1=CC=CC=C1
148.21
148.21
5820-22-4
204°C
C10H12O
MFCD00008597
1933868
LECDNXOCIPRJNJ-UHFFFAOYSA-N
2-methylprop-2-enoxybenzene
79893
96%

Chemical Identifiers

5820-22-4
148.21
LECDNXOCIPRJNJ-UHFFFAOYSA-N
79893
CC(=C)COC1=CC=CC=C1
C10H12O
MFCD00008597
methallyl phenyl ether, 2-methylallyloxy benzene, ether, 2-methylallyl phenyl, benzene, 2-methyl-2-propenyl oxy, 2-methylallyl phenyl ether, 2-methylprop-2-en-1-yl oxy benzene, 2-methyl-2-propen-1-yl oxy benzene, phenyl-methallyl ether, acmc-1b20w
2-methylprop-2-enoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H334-H227-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
May cause an allergic skin reaction.

P210-P261-P272-P280-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P370+P378q-P501c

H227-H317-H334

EINECSNumber : 227-393-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only