Please login to your online account to display your discounted pricing

Methacryloyl chloride, 95%, contains 200 ppm MEHQ as stabilizer, AcroSeal™, ACROS Organics™


Quantity 1L
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC168660010

 View more versions of this product

Catalog No. AC168660010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 104.54
Assay ≥94%
Quantity 1L
Color Clear to slightly colored
Packaging Glass Bottle
TSCA TSCA
RTECS Number OZ5791000
EINECS Number 213-058-9
Density 1.0760g/mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Stabilizer 200ppm MEHQ max.
Infrared Spectrum Authentic
Synonym 2-Methyl-2-propenoyl chloride
Assay Percent Range 95%
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Wear respiratory protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Methacryloyl chloride
Name Note 97%
Linear Formula H2C=C(CH3)COCl
Molecular Formula C4H5ClO
CAS 920-46-7

Specifications

Chemical Name or Material Methacryloyl chloride
Name Note 97%
CAS 920-46-7
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C4H5ClO
Linear Formula H2C=C(CH3)COCl
Formula Weight 104.54
Synonym 2-Methyl-2-propenoyl chloride
Accessories

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Wear respiratory protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.

SDS & Safety Documents