missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methacrylic acid, 99+%, stab. with 250ppm 4-methoxyphenol, Alfa Aesar™

Manufacturer:  Alfa Aesar 15546A1

 View more versions of this product

Catalog No. AA15546A1



Specifications

Specifications

Methacrylic acid
65° (149°F)
1.4310
16°C
2.0 to 2.2
79-41-4
C4H6O2
UN2531
14,5941
CERQOIWHTDAKMF-UHFFFAOYSA-N
2-methylprop-2-enoic acid
4093
86.09
≥99%
1.015g/mL
Acrid
163°C
1kg
Stabilized with 250ppm 4-methoxyphenol
>99%
MFCD00002651
1719937
methacrylic acid, 2-methylacrylic acid, 2-propenoic acid, 2-methyl, methylacrylic acid, 2-methylpropenoic acid, 2-methyl-2-propenoic acid, alpha-methacrylic acid, alpha-methylacrylic acid, 2-methylenepropionic acid, acrylic acid, 2-methyl
CC(=C)C(=O)O
86.09
CHEBI:25219
Liquid

Chemical Identifiers

79-41-4
86.09
CERQOIWHTDAKMF-UHFFFAOYSA-N
4093
2-methylprop-2-enoic acid
C4H6O2
MFCD00002651
methacrylic acid, 2-methylacrylic acid, 2-propenoic acid, 2-methyl, methylacrylic acid, 2-methylpropenoic acid, 2-methyl-2-propenoic acid, alpha-methacrylic acid, alpha-methylacrylic acid, 2-methylenepropionic acid, acrylic acid, 2-methyl
CHEBI:25219
CC(=C)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302-H312-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: METHACRYLIC ACID, STABILIZED

RTECSNumber : OZ2975000

TSCA : TSCA

SDS Documents