missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methacrylic acid, 99%, stab. with 100-250ppm hydroquinone or 4-methoxyphenol

Manufacturer:  Alfa Aesar™ A115430E

 View more versions of this product

Catalog No. AAA115430E



Description

Description

Applications
Methacrylic acid is used as a monomer to prepare polymers. It acts a precursor for the preparation methyl methacrylate, N-substituted methacryl amides, methacrylamide, N-substituted methacryl amides and acryloyl chloride. It enhances the adhesion of surface coatings and adhesives. It acts as auxiliaries for the textile industry. It is involved in the preparation of poly(methacrylic acid)/montmorillonite nanocomposites, which finds application in the leather tanning process.

Solubility
Miscible with warm water, chloroform, ethanol, ether and organic solvents.

Notes
Incompatible with amines, strong bases, strong acids, oxidizing agents and peroxides.
Specifications

Specifications

Methacrylic acid
Acrid
16°C
2.0 to 2.2
79-41-4
MFCD00002651
1719937
methacrylic acid, 2-methylacrylic acid, 2-propenoic acid, 2-methyl, methylacrylic acid, 2-methylpropenoic acid, 2-methyl-2-propenoic acid, alpha-methacrylic acid, alpha-methylacrylic acid, 2-methylenepropionic acid, acrylic acid, 2-methyl
CC(=C)C(=O)O
86.09
CHEBI:25219
99%
65°C (149°F)
163°C
2500g
Stabilized with 100 to 250ppm hydroquinone or 4-methoxyphenol
C4H6O2
UN2531
14,5941
CERQOIWHTDAKMF-UHFFFAOYSA-N
2-methylprop-2-enoic acid
4093
86.09

Chemical Identifiers

79-41-4
86.09
CERQOIWHTDAKMF-UHFFFAOYSA-N
4093
2-methylprop-2-enoic acid
C4H6O2
MFCD00002651
methacrylic acid, 2-methylacrylic acid, 2-propenoic acid, 2-methyl, methylacrylic acid, 2-methylpropenoic acid, 2-methyl-2-propenoic acid, alpha-methacrylic acid, alpha-methylacrylic acid, 2-methylenepropionic acid, acrylic acid, 2-methyl
CHEBI:25219
CC(=C)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302-H312-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: METHACRYLIC ACID, STABILIZED

RTECSNumber : OZ2975000

TSCA : TSCA

SDS Documents