missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methacrylic acid, 99.5%, extra pure, stabilized, ACROS Organics™

Manufacturer:  Acros Organics AC168310251

 View more versions of this product

Catalog No. AC168310251

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methacrylic acid
65°C
Metal Drum
1.01
0.2% max.
Undesignated
25L
79-41-4,150-76-5
99.5%
H2C=C(CH3)CO2H
02, 421
methacrylic acid, 2-methylacrylic acid, 2-propenoic acid, 2-methyl, methylacrylic acid, 2-methylpropenoic acid, 2-methyl-2-propenoic acid, alpha-methacrylic acid, alpha-methylacrylic acid, 2-methylenepropionic acid, acrylic acid, 2-methyl
CC(=C)C(=O)O
86.09
CHEBI:25219
Liquid
1.0100g/mL
Authentic
1.4295 to 1.4315
250ppm (ca.) MEHQ
163.0°C
16.0°C
Stabilized
Extra Pure
C4H6O2
MFCD00002651
15, 6013
CERQOIWHTDAKMF-UHFFFAOYSA-N
2-methylprop-2-enoic acid
4093
86.09
99.5%

Chemical Identifiers

79-41-4
86.09
CERQOIWHTDAKMF-UHFFFAOYSA-N
4093
2-methylprop-2-enoic acid
C4H6O2
MFCD00002651
methacrylic acid, 2-methylacrylic acid, 2-propenoic acid, 2-methyl, methylacrylic acid, 2-methylpropenoic acid, 2-methyl-2-propenoic acid, alpha-methacrylic acid, alpha-methylacrylic acid, 2-methylenepropionic acid, acrylic acid, 2-methyl
CHEBI:25219
CC(=C)C(=O)O
Safety and Handling

Safety and Handling

GHS P Statement
May cause respiratory irritation.
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.

Emergency Overview
Causes severe burns by all exposure routes.
NFPA
Health: 3
Flammability: 1
Instability: 0

SIGNAL WORD: DANGER!

SDS Documents