missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methacrylic acid, 99.5%, extra pure, stabilized, ACROS Organics™

$22.78 - $183.26

Chemical Identifiers

CAS 79-41-4,150-76-5
Assay Percent Range 99.5%
Molecular Formula C4H6O2
Linear Formula H2C=C(CH3)CO2H
Formula Weight 86.09
MDL Number MFCD00002651
Synonym 2-Methylpropenoic acid
Chemical Name or Material Methacrylic acid, Stabilized
Assay ≥99.4%
Grade Extra Pure
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC168310251 SDS Thermo Fisher Scientific
AC168310251
Metal Drum 25L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC168310010 SDS Thermo Fisher Scientific
168310010
Glass Bottle 1L Each for $75.14
AC168310040 SDS Thermo Fisher Scientific
168310040
Plastic Jerrycan 4L Each for $183.26
AC168310250 SDS Thermo Fisher Scientific
168310250
Glass bottle 25mL Each for $22.78
AC168315000 SDS Thermo Fisher Scientific
168315000
Glass bottle 500mL Each for $61.94
AC168312500 SDS Thermo Fisher Scientific
168312500
Glass bottle 250mL Each for $37.65
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

79-41-4,150-76-5
C4H6O2
86.09
2-Methylpropenoic acid
≥99.4%
99.5%
H2C=C(CH3)CO2H
MFCD00002651
Methacrylic acid, Stabilized
Extra Pure

Specifications

86.09
25L
02, 421
Metal Drum
0.2% max.
250ppm (ca.) MEHQ
MFCD00002651
2-Methylpropenoic acid
99.5%
Methacrylic acid, Stabilized
Extra Pure
Liquid
Clear
15, 6013
1.0100g/mL
150-76-5,79-41-4
H2C=C(CH3)CO2H
C4H6O2
1.4295 to 1.4315 (20°C, 589nm)
Authentic
≥99.4%

Safety and Handling

GHS P Statement
May cause respiratory irritation.
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.

Emergency Overview
Causes severe burns by all exposure routes.
NFPA
Health: 3
Flammability: 1
Instability: 0

SIGNAL WORD: DANGER!