Please login to your online account to display your discounted pricing

Methacrylic acid, 99.5%, extra pure, stabilized, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  168312500

 View more versions of this product

Catalog No. AC168312500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.4%
Purity Grade Extra Pure
Physical Form Liquid
Formula Weight 86.09
Quantity 250mL
Color Clear
Beilstein 02, 421
Merck Index 15, 6013
Density 1.0100g/mL
Infrared Spectrum Authentic
Name Note Stabilized
Refractive Index 1.4295 to 1.4315 (20°C, 589nm)
Stabilizer 250ppm (ca.) MEHQ
MDL Number MFCD00002651
CAS 79-41-4
Chemical Name or Material Methacrylic acid
Packaging Glass bottle
Assay Percent Range 99.5%
Molecular Formula C4H6O2
Linear Formula H2C=C(CH3)CO2H
Water 0.2% max.
Synonym 2-Methylpropenoic acid
Health Hazard 1 SIGNAL WORD: DANGER!
CAS 150-76-5
Health Hazard 2 GHS P Statement
May cause respiratory irritation.
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Health Hazard 3 Emergency Overview
Causes severe burns by all exposure routes.
NFPA
Health: 3
Flammability: 1
Instability: 0

Specifications

Chemical Name or Material Methacrylic acid
Name Note Stabilized
CAS 79-41-4
CAS 150-76-5
Purity Grade Extra Pure
Assay ≥99.4%
Assay Percent Range 99.5%
Molecular Formula C4H6O2
Linear Formula H2C=C(CH3)CO2H
Formula Weight 86.09
MDL Number MFCD00002651
Synonym 2-Methylpropenoic acid
Accessories

SIGNAL WORD: DANGER!

Emergency Overview
Causes severe burns by all exposure routes.
NFPA
Health: 3
Flammability: 1
Instability: 0

GHS P Statement
May cause respiratory irritation.
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.

SDS & Safety Documents