missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methacrylic acid, 99.5%, extra pure, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 168310040

 View more versions of this product

Catalog No. AC168310040


 

Specifications Description & Specifications

Specifications

Methacrylic acid
79-41-4,150-76-5
Authentic
1.4295 to 1.4315 (20°C, 589nm)
Liquid
99.5%
C4H6O2
0.2% max.
MFCD00002651
15, 6013
Clear
Stabilized
1.0100g/mL
Plastic Jerrycan
Extra Pure
≥99.4%
250ppm (ca.) MEHQ
H2C=C(CH3)CO2H
86.09
02, 421
2-Methylpropenoic acid
4L

Chemical Identifiers

79-41-4,150-76-5
C4H6O2
86.09
2-Methylpropenoic acid
≥99.4%
99.5%
H2C=C(CH3)CO2H
MFCD00002651
Methacrylic acid, Stabilized
Extra Pure

Safety and Handling

SDS & Safety Documents

GHS P Statement
May cause respiratory irritation.
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.

Emergency Overview
Causes severe burns by all exposure routes.
NFPA
Health: 3
Flammability: 1
Instability: 0

SIGNAL WORD: DANGER!