missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methacrylamide 98.0+%, TCI America™

Manufacturer:  TCI America M0077500G

 View more versions of this product

Catalog No. M0077500G



Specifications

Specifications

Methacrylamide
White
500g
C4H7NO
methacrylamide, 2-methylacrylamide, methacrylic amide, methacrylic acid amide, 2-propenamide, 2-methyl, 2-methylpropenamide, methacryamide, methylacrylic amide, mhoromer bm801, alpha-methyl acrylic amide
CC(=C)C(=O)N
85.106
CHEBI:51759
Crystal-Powder at 20°C
215°C
110°C
79-39-0
MFCD00008018
FQPSGWSUVKBHSU-UHFFFAOYSA-N
2-methylprop-2-enamide
6595
85.11
≥98.0% (GC,T)

Chemical Identifiers

79-39-0
85.106
FQPSGWSUVKBHSU-UHFFFAOYSA-N
6595
2-methylprop-2-enamide
C4H7NO
MFCD00008018
methacrylamide, 2-methylacrylamide, methacrylic amide, methacrylic acid amide, 2-propenamide, 2-methyl, 2-methylpropenamide, methacryamide, methylacrylic amide, mhoromer bm801, alpha-methyl acrylic amide
CHEBI:51759
CC(=C)C(=O)N
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1065

RTECSNumber : UC6475000

TSCA : Yes

Documents