missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methacholine Bromide 99.0+%, TCI America™
$175.86 - $527.56
Chemical Identifiers
CAS | 333-31-3 |
---|---|
Molecular Formula | C8H18BrNO2 |
Molecular Weight (g/mol) | 240.14 |
MDL Number | MFCD00011816 |
InChI Key | MMVPLEUBMWUYIB-UHFFFAOYNA-M |
Synonym | methacholine bromide, 2-acetoxy-n,n,n-trimethylpropan-1-aminium bromide, acetyl beta-methylcholine bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide 1:1, methacholini bromidum, methacholine bromide nf, c8h18no2.br, mecholyl bromide, methacholinebromide |
PubChem CID | 92754 |
IUPAC Name | [2-(acetyloxy)propyl]trimethylazanium bromide |
SMILES | [Br-].CC(C[N+](C)(C)C)OC(C)=O |
Chemical Identifiers
333-31-3 | |
240.14 | |
MMVPLEUBMWUYIB-UHFFFAOYNA-M | |
92754 | |
[Br-].CC(C[N+](C)(C)C)OC(C)=O |
C8H18BrNO2 | |
MFCD00011816 | |
methacholine bromide, 2-acetoxy-n,n,n-trimethylpropan-1-aminium bromide, acetyl beta-methylcholine bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide 1:1, methacholini bromidum, methacholine bromide nf, c8h18no2.br, mecholyl bromide, methacholinebromide | |
[2-(acetyloxy)propyl]trimethylazanium bromide |
Specifications
333-31-3 | |
White | |
MFCD00011816 | |
methacholine bromide, 2-acetoxy-n,n,n-trimethylpropan-1-aminium bromide, acetyl beta-methylcholine bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide, 1-propanaminium, 2-acetyloxy-n,n,n-trimethyl-, bromide 1:1, methacholini bromidum, methacholine bromide nf, c8h18no2.br, mecholyl bromide, methacholinebromide | |
[Br-].CC(C[N+](C)(C)C)OC(C)=O | |
240.14 | |
240.14 | |
Crystalline Powder |
147°C | |
C8H18BrNO2 | |
25 g | |
MMVPLEUBMWUYIB-UHFFFAOYNA-M | |
[2-(acetyloxy)propyl]trimethylazanium bromide | |
92754 | |
≥99.0% (T) | |
Methacholine Bromide |
Safety and Handling
TSCA : Yes