Please login to your online account to display your discounted pricing

Metergoline, Tocris

5-HT2 antagonist. Also 5-HT1 antagonist and 5-HT1D ligand; Has moderate affinity for 5-HT6 and high affinity for 5-HT7

$88.05 - $323.08

Chemical Identifiers

Chemical Name or Material Metergoline
Assay Percent Range >98%
Molecular Formula C25H29N3O2
Formula Weight 403.52
Synonym [(8β)-1, 6-Dimethylergolin-8-yl)methyl]carbamic acid phenylmethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

059010

 
r & d systems
RDS059010
10mg Each for $88.05

059050

 
r & d systems
RDS059050
50mg Each for $323.08
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 403.52
Assay Percent Range >98%
Recommended Storage Store at Room Temperature
Synonym [(8β)-1, 6-Dimethylergolin-8-yl)methyl]carbamic acid phenylmethyl ester
Molecular Formula C25H29N3O2
CAS 17692-51-2
Chemical Name or Material Metergoline
Safety and Handling

Store at Room Temperature