Please login to your online account to display your discounted pricing

Metergoline, Tocris


5-HT2 antagonist. Also 5-HT1 antagonist and 5-HT1D ligand; Has moderate affinity for 5-HT6 and high affinity for 5-HT7

Quantity 50mg

Manufacturer: r & d systems  RDS059050

 View more versions of this product

Catalog No. 059050

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 403.52
Molecular Formula C25H29N3O2
Synonym [(8β)-1, 6-Dimethylergolin-8-yl)methyl]carbamic acid phenylmethyl ester
CAS 17692-51-2
Chemical Name or Material Metergoline
Assay Percent Range >98%
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Metergoline
Assay Percent Range >98%
Molecular Formula C25H29N3O2
Formula Weight 403.52
Synonym [(8β)-1, 6-Dimethylergolin-8-yl)methyl]carbamic acid phenylmethyl ester
Accessories

Store at Room Temperature