missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Metaxalone, TCI America™

$76.39 - $256.67

Chemical Identifiers

CAS 1665-48-1
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.256
MDL Number MFCD00867700
InChI Key IMWZZHHPURKASS-UHFFFAOYSA-N
Synonym 5-(3,5-Dimethylphenoxymethyl)-2-oxazolidinone, 5-[(3,5-Xylyloxy)methyl]-2-oxazolidinone
PubChem CID 15459
ChEBI CHEBI:6797
IUPAC Name 5-[(3,5-dimethylphenoxy)methyl]-1,3-oxazolidin-2-one
SMILES CC1=CC(=CC(=C1)OCC2CNC(=O)O2)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25781G View Documents TCI America
M25781G
1g Each for $76.39
 
M25785G View Documents TCI America
M25785G
5g Each for $256.67
 
Specifications

Chemical Identifiers

1665-48-1
221.256
IMWZZHHPURKASS-UHFFFAOYSA-N
15459
5-[(3,5-dimethylphenoxy)methyl]-1,3-oxazolidin-2-one
C12H15NO3
MFCD00867700
5-(3,5-Dimethylphenoxymethyl)-2-oxazolidinone, 5-[(3,5-Xylyloxy)methyl]-2-oxazolidinone
CHEBI:6797
CC1=CC(=CC(=C1)OCC2CNC(=O)O2)C

Specifications

225°C
124°C
1665-48-1
MFCD00867700
IMWZZHHPURKASS-UHFFFAOYSA-N
5-[(3,5-dimethylphenoxy)methyl]-1,3-oxazolidin-2-one
15459
221.26
≥98.0% (GC)
White
1g
C12H15NO3
5-(3,5-Dimethylphenoxymethyl)-2-oxazolidinone, 5-[(3,5-Xylyloxy)methyl]-2-oxazolidinone
CC1=CC(=CC(=C1)OCC2CNC(=O)O2)C
221.256
CHEBI:6797
Crystal-Powder at 20°C
Metaxalone
Safety and Handling

Safety and Handling

RTECSNumber : RQ3850000

TSCA : No

Recommended Storage : Freezer