Learn More
Metaphosphoric acid, stabilized, Thermo Scientific Chemicals
Metaphosphoric acid, 40 to 46% (HPO3), HO3P, CAS Number-37267-86-0, 50813-16-6, hydrogenphosphonate, metaphosphoric acid, meta-phosphoric acid, hydrogen phosphonate, phosphenic acid, m-phosphoric acid, metaphosphoric acid hpo3, phosphonate 1-, x-phosphate-group, unii-mtk99r3uv0, 500g, 5ppm max.
Supplier: Thermo Scientific Chemicals 219225000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Metaphosphoric acid | |
50ppm max. | |
500 g | |
Glassy Lump | |
MFCD00011351 | |
15, 7457 | |
Solubility in water: soluble. Other solubilities: slowly forming the ortho-acid, soluble in alcohol | |
OP(=O)=O | |
3084658 | |
40 to 46% (HPO3) |
37267-86-0,50813-16-6 | |
Glass bottle | |
HO3P | |
(HPO3)n | |
01,665; 16,177 | |
metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate | |
UEZVMMHDMIWARA-UHFFFAOYSA-N | |
79.979 | |
79.98 |
Chemical Identifiers
37267-86-0 | |
79.979 | |
UEZVMMHDMIWARA-UHFFFAOYSA-N | |
3084658 |
HO3P | |
MFCD00011351 | |
metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate | |
OP(=O)=O |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 253-433-4
TSCA : TSCA
RUO – Research Use Only