Metaphosphoric acid, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 450251000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Metaphosphoric acid | |
Glass Bottle | |
HO3P | |
metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate | |
UEZVMMHDMIWARA-UHFFFAOYSA-N | |
79.979 | |
ACS Reagent |
37267-86-0,50813-16-6 | |
100 g | |
(HPO3)n | |
Solubility in water: soluble. Other solubilities: slowly forming the ortho-acid, soluble in alcohol | |
OP(=O)=O | |
3084658 |
Chemical Identifiers
37267-86-0 | |
79.979 | |
metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate | |
OP(=O)=O |
HO3P | |
UEZVMMHDMIWARA-UHFFFAOYSA-N | |
3084658 |
RUO – Research Use Only