Please login to your online account to display your discounted pricing

Metaphit, Tocris


Acylator of PCP and σ receptors

Quantity 10mg

Manufacturer: r & d systems  RDS056810

 View more versions of this product

Catalog No. 056810

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 300.46
Assay Percent Range >98%
Chemical Name or Material Metaphit
Molecular Formula C18H24N2S
Synonym 1-(1-(3-Isothiocyanato)phenyl)cyclohexylpiperidine
CAS 96316-00-6
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material Metaphit
Assay Percent Range >98%
Molecular Formula C18H24N2S
Formula Weight 300.46
Synonym 1-(1-(3-Isothiocyanato)phenyl)cyclohexylpiperidine
Accessories

Desiccate at -20°C