Please login to your online account to display your discounted pricing

Metaphit, Tocris

Acylator of PCP and σ receptors

$129.05 - $546.19

Chemical Identifiers

Chemical Name or Material Metaphit
Assay Percent Range >98%
Molecular Formula C18H24N2S
Formula Weight 300.46
Synonym 1-(1-(3-Isothiocyanato)phenyl)cyclohexylpiperidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

056850

 
r & d systems
RDS056850
50mg Each for $546.19

056810

 
r & d systems
RDS056810
10mg Each for $129.05
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 300.46
Molecular Formula C18H24N2S
Synonym 1-(1-(3-Isothiocyanato)phenyl)cyclohexylpiperidine
Recommended Storage Desiccate at -20°C
Chemical Name or Material Metaphit
Assay Percent Range >98%
CAS 96316-00-6
Safety and Handling

Desiccate at -20°C