missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Metanilic acid, 98+%

Manufacturer:  Alfa Aesar™ A1457422

 View more versions of this product

Catalog No. AAA1457422



Description

Description

Applications
Metanilic acid was used in the synthesis of cytocompatible sulfonated polyanilines. It was used in fabrication of novel glucose biosensor having large active surface area and excellent conductivity. It undergoes microbial desulfonation to 3-aminophenol by Pseudomonas sp. strain S-313. It can be used to produce CN3O6(1-)*Cs(1+) and dicesium salt of N-nitrometanilic acid at temperature of 25°C.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
Specifications

Specifications

Metanilic acid
100g
C6H7NO3S
473264
metanilic acid, m-sulfanilic acid, m-aminobenzenesulfonic acid, m-anilinesulfonic acid, 1-aminobenzene-3-sulfonic acid, benzenesulfonic acid, 3-amino, aminobenzenesulfonic acid, kyselina metanilova, 3-amino-benzenesulfonic acid, 3-sulfoaniline
C1=CC(=CC(=C1)S(=O)(=O)O)N
173.186
CHEBI:27764
≥98%
>300°C
121-47-1
MFCD00065345
14,5927
ZAJAQTYSTDTMCU-UHFFFAOYSA-N
3-aminobenzenesulfonic acid
8474
173.19

Chemical Identifiers

121-47-1
173.186
ZAJAQTYSTDTMCU-UHFFFAOYSA-N
8474
3-aminobenzenesulfonic acid
C6H7NO3S
MFCD00065345
metanilic acid, m-sulfanilic acid, m-aminobenzenesulfonic acid, m-anilinesulfonic acid, 1-aminobenzene-3-sulfonic acid, benzenesulfonic acid, 3-amino, aminobenzenesulfonic acid, kyselina metanilova, 3-amino-benzenesulfonic acid, 3-sulfoaniline
CHEBI:27764
C1=CC(=CC(=C1)S(=O)(=O)O)N
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : OY2300000

TSCA : TSCA

Documents