Please login to your online account to display your discounted pricing

Alfa Aesar™ Metanilic acid, 98+%

$31.86 - $116.22

Chemical Identifiers

Chemical Name or Material Metanilic acid
CAS 121-47-1
Molecular Formula C6H7NO3S
Formula Weight 173.19
MDL Number MFCD00065345
Beilstein 473264
Merck Index 14,5927
Synonym 3-Aminobenzenesulfonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1457422

  SDS
alfa aesar™
A1457422
100g Each for $31.86

AAA1457436

  SDS
alfa aesar™
A1457436
500g Each for $116.22
Description & Specifications

Specifications

Formula Weight 173.19
Chemical Name or Material Metanilic acid
TSCA TSCA
Assay ≥98%
Molecular Formula C6H7NO3S
MDL Number MFCD00065345
CAS 121-47-1
Beilstein 473264
Merck Index 14,5927
Synonym 3-Aminobenzenesulfonic acid
Health Hazard 1 Warning
Melting Point >300°C
Quantity 100g
Health Hazard 2 GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number OY2300000

Applications
Metanilic acid was used in the synthesis of cytocompatible sulfonated polyanilines. It was used in fabrication of novel glucose biosensor having large active surface area and excellent conductivity. It undergoes microbial desulfonation to 3-aminophenol by Pseudomonas sp. strain S-313. It can be used to produce CN3O6(1-)*Cs(1+) and dicesium salt of N-nitrometanilic acid at temperature of 25°C.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.