missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Metacresol Purple, TCI America™

Manufacturer:  TCI America M007425G

 View more versions of this product

Catalog No. M007425G



Specifications

Specifications

Metacresol Purple
C21H18O5S
m-Cresol Purple
CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C
382.43
382.43
Undesignated
2303-01-7
MFCD00005871
OLQIKGSZDTXODA-UHFFFAOYSA-N
4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
73030
Crystal-Powder at 20°C
25g

Chemical Identifiers

2303-01-7
382.43
OLQIKGSZDTXODA-UHFFFAOYSA-N
73030
CC1=C(C=CC(=C1)O)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=C(C=C(C=C4)O)C
C21H18O5S
MFCD00005871
m-Cresol Purple
4-[3-(4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
Safety and Handling

Safety and Handling

TSCA : Yes

Documents