Please login to your online account to display your discounted pricing

meta-Phosphoric Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$1,046.49 - $3,559.78

Chemical Identifiers

Chemical Name or Material meta-Phosphoric Acid
CAS 37267-86-0
Purity Grade Baker Analyzed™ ACS Reagent
Purity Grade Notes Meets ACS reagent chemical requirements
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003332

 
avantor performance
025205
Poly Bottle 2.5kg Each for $3,559.78

02003333

 
avantor performance
025201
Poly Bottle 500g Each for $1,046.49
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Purity Grade Notes Meets ACS reagent chemical requirements
Chemical Name or Material meta-Phosphoric Acid
Specific Gravity 1L = 0.00kg
CAS 37267-86-0
Quantity 2.5kg
Packaging Poly Bottle